Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002775.4 | NP_002766.1 | 2 | 690 | ||
GenBank mRNA | AF097709.1 | - | 2 | 133 | ||
AK091944.1 | - | 4 | 465 | |||
AK092476.1 | - | 3 | 471 | |||
AK290089.1 | - | 2 | 181 | |||
BC011352.1 | - | 3 | 457 | |||
BC031082.1 | - | 2 | 598 | |||
D87258.1 | - | 2 | 610 | |||
DA633502.1 | - | 4 | 465 | |||
Y07921.1 | - | 2 | 600 |