Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146754.2 | NP_666965.2 | 1 | 628 | ||
GenBank mRNA | BC138388.1 | - | 1 | 628 | 8861408 |