Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146758.1 | NP_666969.1 | 1 | 895 | ||
GenBank mRNA | BC128570.1 | - | 1 | 897 | 40135306 |