Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146764.1 | NP_666975.1 | 1 | 594 | ||
GenBank mRNA | BC119111.1 | - | 1 | 615 | 8733860 | |
BC120665.1 | - | 1 | 615 | 40129588 |