Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146773.1 | NP_666984.1 | 1 | 136 | ||
GenBank mRNA | BC130241.1 | - | 1 | 140 | 40142176 |