Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146774.1 | NP_666985.1 | 1 | 235 | ||
GenBank mRNA | BC138079.1 | - | 1 | 327 | 8861098 | |
BC145756.1 | - | 1 | 327 | 40131059 |
Gene Symbol: Olfr473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC148218.1 | - | 1 | 239 | 40134717 |