Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042465.2 | NP_001035930.1 | 6, 7 | 877 | ||
NM_001042466.2 | NP_001035931.1 | 6, 7 | 877 | |||
NM_002778.3 | NP_002769.1 | 6, 7 | 877 | |||
GenBank mRNA | AB209776.1 | - | 6, 7 | 732 | ||
AK057878.1 | - | 6, 7 | 641 | |||
AK129790.1 | - | 6, 7 | 753 | |||
AK223290.1 | - | 6, 7 | 751 | |||
AK293672.1 | - | 7, 8 | 692 | |||
AK299184.1 | - | 6, 7 | 787 | |||
BC001503.2 | - | 6, 7 | 728 | 3508263 | ||
BC004275.1 | - | 6, 7 | 736 | 3615413 | ||
BC007612.1 | - | 6, 7 | 741 | 3351606 | ||
BT006849.1 | - | 6, 7 | 723 | |||
CR456746.1 | - | 6, 7 | 723 | |||
D00422.1 | - | 6, 7 | 731 | |||
J03015.1 | - | 6, 7 | 729 | |||
J03077.1 | - | 6, 7 | 761 | |||
M12710.1 | - | 1, 2 | 141 | |||
M32221.1 | - | 6, 7 | 729 | |||
M81355.1 | - | 6, 7 | 731 |