Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184825.1 | NP_001171754.1 | 2 | 515 | ||
NM_001184826.1 | NP_001171755.1 | 2 | 515 | |||
NM_001297773.1 | NP_001284702.1 | 2 | 463 | |||
NM_006905.2 | NP_008836.2 | 2 | 515 | |||
XM_005259065.3 | XP_005259122.1 | 2 | 514 | |||
XM_011527125.1 | XP_011525427.1 | 2 | 514 | |||
XM_017026994.1 | XP_016882483.1 | 2 | 514 | |||
GenBank mRNA | BC022338.1 | - | 2 | 515 | 4732357 | |
BC058285.1 | - | 2 | 439 | 4770394 | ||
CR450329.1 | - | 2 | 377 | |||
J04539.1 | - | 2 | 437 | |||
M17908.1 | - | 2 | 443 | |||
M20879.1 | - | 2 | 463 | |||
M20881.1 | - | 2 | 494 | |||
M21822.1 | - | 2 | 450 | |||
M25385.1 | - | 2 | 413 | |||
M33663.1 | - | 2 | 440 | |||
M33664.1 | - | 2 | 377 | |||
M34715.1 | - | 2 | 538 | |||
M37397.1 | - | 2 | 467 |
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC008405.1 | - | 2 | 474 | 4043484 |