Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297773.1 | NP_001284702.1 | 6 | 1369 | ||
NM_006905.2 | NP_008836.2 | 5 | 1776 | |||
XM_005259065.3 | XP_005259122.1 | 4 | 1496 | |||
XM_011527125.1 | XP_011525427.1 | 4 | 1496 | |||
GenBank mRNA | BC022338.1 | - | 5 | 1776 | 4732357 | |
M20879.1 | - | 6 | 1369 | |||
M34715.1 | - | 5 | 1799 |
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026995.1 | XP_016882484.1 | 6 | 1939 | ||
XM_017026998.1 | XP_016882487.1 | 5 | 1664 | |||
XM_017026999.1 | XP_016882488.1 | 5 | 1665 | |||
XM_017027001.1 | XP_016882490.1 | 4 | 1491 | |||
XM_017027002.1 | XP_016882491.1 | 4 | 1491 |