Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000021.3 | NP_000012.1 | 12 | 1886 | ||
NM_007318.2 | NP_015557.2 | 12 | 1874 | |||
XM_005267864.2 | XP_005267921.1 | 12 | 1880 | |||
XM_005267866.1 | XP_005267923.1 | 12 | 1869 | |||
XM_011536971.2 | XP_011535273.1 | 12 | 1778 | |||
XM_011536972.2 | XP_011535274.1 | 12 | 2142 | |||
XM_011536973.1 | XP_011535275.1 | 12 | 2130 | |||
XM_011536974.1 | XP_011535276.1 | 12 | 1771 | |||
GenBank mRNA | AK122722.1 | - | 10 | 3155 | ||
AK303876.1 | - | 8 | 1261 | |||
BC011729.2 | - | 12 | 1855 | 4423098 | ||
L42110.1 | - | 12 | 1850 | |||
L76517.1 | - | 12 | 2143 |