Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276495.1 | NP_001263424.1 | 4 | 1048 | ||
NM_001316339.1 | NP_001303268.1 | 4 | 961 | |||
NM_002780.4 | NP_002771.2 | 5 | 1327 | |||
NM_213633.2 | NP_998798.1 | 4 | 1048 | |||
XM_017026995.1 | XP_016882484.1 | 6 | 1390 | |||
XM_017026996.1 | XP_016882485.1 | 6 | 1389 | |||
XM_017026997.1 | XP_016882486.1 | 5 | 1221 | |||
XM_017026998.1 | XP_016882487.1 | 5 | 1115 | |||
XM_017026999.1 | XP_016882488.1 | 5 | 1116 | |||
XM_017027000.1 | XP_016882489.1 | 5 | 1114 | |||
XM_017027001.1 | XP_016882490.1 | 4 | 942 | |||
XM_017027002.1 | XP_016882491.1 | 4 | 942 | |||
GenBank mRNA | AF289606.1 | - | 5 | 668 | ||
AK075149.1 | - | 5 | 942 | |||
BC008405.1 | - | 5, 6 | 1186 | 4043484 | ||
BC063127.1 | - | 5 | 1195 | 4768860 | ||
M33665.1 | - | 3 | 468 | |||
M94891.1 | - | 5 | 1175 | |||
U18468.1 | - | 5 | 1094 | |||
U18469.1 | - | 4 | 837 | |||
X17097.1 | - | 5 | 1181 |