Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276495.1 | NP_001263424.1 | 4 | 1125 | ||
NM_001316339.1 | NP_001303268.1 | 4 | 1038 | |||
NM_002780.4 | NP_002771.2 | 5 | 1404 | |||
NM_213633.2 | NP_998798.1 | 4 | 1125 | |||
XM_017026995.1 | XP_016882484.1 | 6 | 1467 | |||
XM_017026996.1 | XP_016882485.1 | 6 | 1466 | |||
XM_017026997.1 | XP_016882486.1 | 5 | 1298 | |||
XM_017026998.1 | XP_016882487.1 | 5 | 1192 | |||
XM_017026999.1 | XP_016882488.1 | 5 | 1193 | |||
XM_017027000.1 | XP_016882489.1 | 5 | 1191 | |||
XM_017027001.1 | XP_016882490.1 | 4 | 1019 | |||
XM_017027002.1 | XP_016882491.1 | 4 | 1019 | |||
GenBank mRNA | AF289606.1 | - | 5 | 745 | ||
AK075149.1 | - | 5 | 1019 | |||
BC008405.1 | - | 6 | 1263 | 4043484 | ||
BC063127.1 | - | 5 | 1272 | 4768860 | ||
M33665.1 | - | 3 | 545 | |||
M94891.1 | - | 5 | 1252 | |||
U18468.1 | - | 5 | 1171 | |||
U18469.1 | - | 4 | 914 | |||
X17097.1 | - | 5 | 1258 |