Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130014.1 | NP_001123486.1 | 2 | 347 | ||
NM_002781.3 | NP_002772.3 | 2 | 347 | |||
XM_011527132.2 | XP_011525434.1 | 2 | 345 | |||
XM_017027003.1 | XP_016882492.1 | 2 | 345 | |||
GenBank mRNA | BC012607.1 | - | 2 | 314 | 4043912 | |
BG622527.1 | - | 2 | 315 | |||
DA835167.1 | - | 2 | 347 | |||
M25384.1 | - | 2 | 315 | |||
M73713.1 | - | 2 | 301 |