Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146871.2 | NP_667082.2 | 1 | 702 | ||
GenBank mRNA | BC127995.1 | - | 1 | 706 | 40135758 |
Gene Symbol: Olfr251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207553.2 | NP_997436.2 | 1 | 702 |
Gene Symbol: Olfr900 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146874.1 | NP_667085.1 | 1 | 702 | ||
GenBank mRNA | BC106986.2 | - | 1 | 702 | 40043276 |