Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113410.1 | NP_001106881.1 | 3 | 483 | ||
NM_002785.2 | NP_002776.3 | 4 | 849 | |||
NM_203287.1 | NP_976032.2 | 3 | 483 | |||
GenBank mRNA | BC020711.1 | - | 3 | 473 | 4732286 | |
BG571293.1 | - | 2 | 192 | |||
M69245.1 | - | 4 | 811 | |||
R66665.1 | - | 2 | 197 | |||
U25988.1 | - | 3 | 479 |