Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr870 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146904.1 | NP_667115.1 | 1 | 444 |
Gene Symbol: Olfr871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146903.2 | NP_667114.2 | 1 | 588 | ||
GenBank mRNA | BC119201.1 | - | 1 | 494 | 8733950 | |
BC119203.1 | - | 1 | 494 | 8733952 |