Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146908.1 | NP_667119.1 | 1 | 662 | ||
GenBank mRNA | BC119051.1 | - | 1 | 705 | 8733800 | |
BC119053.1 | - | 1 | 705 | 8733802 |