Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146923.2 | NP_667134.2 | 2 | 711 | ||
GenBank mRNA | AY053400.1 | - | 1 | 215 | ||
BC138172.1 | - | 1 | 614 | 8861191 | ||
BC138173.1 | - | 1 | 614 | 8861192 | ||
X89679.1 | - | 1 | 220 | |||
X89692.1 | - | 1 | 220 |
Gene Symbol: Olfr376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172686.1 | NP_001166157.1 | 3 | 859 | ||
NM_146922.2 | NP_667133.1 | 2 | 728 |