Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr353 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146941.1 | NP_667152.1 | 1 | 281 |
Gene Symbol: Olfr351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146942.1 | NP_667153.1 | 1 | 281 |