Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146962.1 | NP_667173.1 | 1 | 68 | ||
GenBank mRNA | BC119150.1 | - | 1 | 156 | 8733899 | |
BC120836.1 | - | 1 | 156 | 40129759 |
Gene Symbol: Olfr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146963.1 | NP_667174.1 | 1 | 65 | ||
GenBank mRNA | BC125298.1 | - | 1 | 88 | 40129813 | |
BC125300.1 | - | 1 | 88 | 40129815 |