Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146973.2 | NP_667184.2 | 1 | 296 | ||
GenBank mRNA | BC104287.1 | - | 1 | 296 | ||
BC104288.1 | - | 1 | 296 | |||
BC106983.1 | - | 1 | 296 | |||
BC127132.1 | - | 1 | 296 | 40042541 | ||
BC127133.1 | - | 1 | 296 | 40042542 |