Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146341.1 | NP_666453.1 | 1 | 637 |
Gene Symbol: Olfr1255 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146977.2 | NP_667188.2 | 1 | 655 |