Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002793.3 | NP_002784.1 | 6 | 760 | ||
GenBank mRNA | AB451312.1 | - | 6 | 673 | ||
AB451442.1 | - | 6 | 673 | |||
AK023290.1 | - | 5 | 1888 | |||
AK058050.1 | - | 6 | 730 | |||
AK223193.1 | - | 6 | 711 | |||
BC000508.2 | - | 6 | 711 | 2822268 | ||
BC020807.1 | - | 6 | 711 | 4281426 | ||
BQ685374.1 | - | 5 | 490 | |||
BT019720.1 | - | 6 | 673 | |||
D00761.1 | - | 6 | 685 |