Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270481.1 | NP_001257410.1 | 5 | 551 | ||
NM_002798.2 | NP_002789.1 | 5 | 551 | |||
GenBank mRNA | AK311768.1 | - | 5 | 514 | ||
BC000835.2 | - | 5 | 518 | 3456009 | ||
BF671774.1 | - | 5 | 549 | |||
BM553399.1 | - | 5 | 498 | |||
BQ691763.1 | - | 5 | 533 | |||
BT009809.1 | - | 5 | 500 | |||
BU848746.1 | - | 5 | 483 | |||
CR457065.1 | - | 5 | 500 | |||
D29012.1 | - | 5 | 509 | |||
X61971.1 | - | 3 | 323 |