Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147056.1 | NP_667267.1 | 1 | 384 | ||
GenBank mRNA | BC127980.1 | - | 1 | 388 | 40135216 |