Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr691 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147061.1 | NP_667272.1 | 1 | 467 | ||
GenBank mRNA | BC104116.2 | - | 1 | 459 | ||
BC104117.1 | - | 1 | 459 | |||
BC119417.2 | - | 1 | 467 | 40044083 | ||
BC119418.1 | - | 1 | 467 | 40044084 |