Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147068.1 | NP_667279.1 | 1 | 764 | ||
GenBank mRNA | BC116856.1 | - | 1 | 811 | 40126175 | |
BC119189.1 | - | 1 | 811 | 8733938 |
Gene Symbol: Olfr168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146357.1 | NP_666469.1 | 1 | 764 | ||
GenBank mRNA | BC127969.1 | - | 1 | 768 | 40135070 |