Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002802.2 | NP_002793.2 | 11 | 1281 | ||
XM_017021470.1 | XP_016876959.1 | 11 | 2800 | |||
XM_017021471.1 | XP_016876960.1 | 12 | 1357 | |||
GenBank mRNA | AB451311.1 | - | 1 | 1233 | ||
AB451441.1 | - | 1 | 1233 | |||
AK124201.1 | - | 3 | 1941 | |||
AK222668.1 | - | 11 | 1249 | |||
AK299121.1 | - | 10 | 1157 | |||
AK313325.1 | - | 11 | 1270 | |||
BC000512.2 | - | 1 | 1267 | 2822718 | ||
BC016368.1 | - | 1 | 1261 | 4133348 | ||
BC018859.2 | - | 1 | 1234 | |||
BC067741.1 | - | 1 | 1254 | 5264945 | ||
BC073818.1 | - | 1 | 1239 | 5456334 | ||
BT009826.1 | - | 1 | 1233 | |||
CR457044.1 | - | 1 | 1233 | |||
L02426.1 | - | 11 | 1293 |