Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147105.2 | NP_667316.2 | 1 | 739 | ||
GenBank mRNA | BC114537.1 | - | 1 | 738 |
Gene Symbol: Olfr979 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147108.2 | NP_667319.1 | 1 | 764 | ||
GenBank mRNA | BC116754.1 | - | 1 | 764 | 40126073 | |
BC116756.1 | - | 1 | 764 | 40126075 |
Gene Symbol: Olfr980 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147106.2 | NP_667317.1 | 1 | 821 | ||
GenBank mRNA | BC116814.1 | - | 1 | 821 | 40126133 | |
BC119187.1 | - | 1 | 821 | 8733936 |