Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006503.3 | NP_006494.1 | 11 | 1396 | ||
NM_153001.2 | NP_694546.1 | 11 | 1303 | |||
GenBank mRNA | AF020736.1 | - | 11 | 1238 | ||
AF038965.1 | - | 11 | 1220 | |||
AK290285.1 | - | 11 | 1204 | |||
AK313183.1 | - | 11 | 1238 | |||
BC000343.2 | - | 11 | 1218 | 2823007 | ||
BC010396.1 | - | 11 | 1142 | 4046205 | ||
BC014488.1 | - | 11 | 1218 | 4899042 | ||
BC018811.2 | - | 11 | 1218 | |||
BT007232.1 | - | 11 | 1198 | |||
U27515.1 | - | 11 | 1210 |