Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190313.1 | NP_001177242.1 | 1 | 168 | ||
NM_145613.4 | NP_663588.2 | 1 | 168 | |||
GenBank mRNA | AF469650.1 | - | 1 | 96 | ||
AK053629.1 | - | 1 | 169 |
Gene Symbol: Mfrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190314.1 | NP_001177243.1 | 1 | 168 | ||
NM_147126.3 | NP_667337.1 | 1 | 168 |