Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153804.4 | NP_722499.4 | 19 | 4279 | ||
GenBank mRNA | AK144041.1 | - | 2690 | |||
AK145222.1 | - | 2 | 2085 | |||
AK172977.1 | - | 5 | 4965 | |||
BC031136.1 | - | 3 | 2300 | 4971548 | ||
BC096443.1 | - | 19 | 4257 | 4189193 |