Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153804.4 | NP_722499.4 | 18 | 2891 | ||
GenBank mRNA | AK144041.1 | - | 1302 | |||
AK172977.1 | - | 4 | 3577 | |||
BC031136.1 | - | 2 | 912 | 4971548 | ||
BC096443.1 | - | 18 | 2869 | 4189193 |