Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002808.4 | NP_002799.3 | 3 | 420 | ||
GenBank mRNA | AB209843.1 | - | 3 | 266 | ||
AK095245.1 | - | 3 | 811 | |||
AK297256.1 | - | 4 | 251 | |||
AK315474.1 | - | 3 | 279 | |||
AL162014.1 | - | 3 | 283 | |||
BC002368.2 | - | 3 | 252 | 2822191 | ||
BC002997.1 | - | 3 | 273 | 2905429 | ||
BC007897.2 | - | 3 | 251 | 4131391 | ||
BT009736.1 | - | 3 | 251 | |||
D78151.1 | - | 3 | 284 | |||
HY006627.1 | - | 3 | 421 | |||
U12596.1 | - | 3 | 192 | |||
X86446.1 | - | 3 | 224 |