Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170756.2 | NP_739562.1 | 3 | 1189 | ||
GenBank mRNA | AK051741.1 | - | 3 | 1135 | ||
AK122364.1 | - | 1 | 1831 | |||
AK144826.1 | - | 3 | 1161 | |||
AK156188.1 | - | 3 | 1190 | |||
AK162743.1 | - | 3 | 1189 | |||
AK170356.1 | - | 3 | 1190 | |||
AK171240.1 | - | 3 | 1163 | |||
AK171664.1 | - | 3 | 1167 | |||
AK171852.1 | - | 3 | 1179 | |||
AK191027.1 | - | 1 | 97 | |||
AK194166.1 | - | 1 | 97 | |||
BC034597.1 | - | 3 | 1178 | 5363513 |