Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153680.2 | NP_710147.1 | 6 | 589 | ||
GenBank mRNA | AK047265.1 | - | 5 | 683 | ||
AK077650.1 | - | 6 | 591 | |||
AK077780.1 | - | 6 | 591 | |||
AK156299.1 | - | 6 | 640 | |||
AK219724.1 | - | 3 | 234 | |||
BC023732.1 | - | 6 | 605 | 5352355 | ||
BC023925.1 | - | 1 | 52 | |||
BC026571.1 | - | 6 | 589 | 4986268 | ||
BC029675.1 | - | 6 | 576 |