Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153680.2 | NP_710147.1 | 7 | 686 | ||
GenBank mRNA | AK047265.1 | - | 6 | 780 | ||
AK077650.1 | - | 7 | 688 | |||
AK156299.1 | - | 7 | 737 | |||
AK219724.1 | - | 4 | 331 | |||
BC023732.1 | - | 7 | 702 | 5352355 | ||
BC023925.1 | - | 2 | 149 | |||
BC026571.1 | - | 7 | 686 | 4986268 | ||
BC029675.1 | - | 7 | 673 |