Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013766.2 | NP_001013788.2 | 1, 2 | 110 | ||
NM_001115154.1 | NP_001108626.1 | 1, 2 | 110 | |||
GenBank mRNA | AK082201.1 | - | 1, 2 | 111 | ||
BC150876.1 | - | 1, 2 | 111 | 9087787 |