Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam19a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252387.1 | NP_001239316.1 | 3, 4 | 1734 | ||
NM_182807.3 | NP_877959.2 | 4, 5 | 1946 | |||
GenBank mRNA | AK032077.1 | - | 1, 2 | 50 | ||
AK032483.1 | - | 3, 4 | 816 | |||
AK139403.1 | - | 3, 4 | 1818 | |||
AK140763.1 | - | 3, 4 | 1697 | |||
AK164201.1 | - | 3, 4 | 826 | |||
AY325121.1 | - | 4, 5 | 633 | |||
BC027082.1 | - | 4, 5 | 1832 | 5360443 |