Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290654.1 | NP_001277583.1 | 9 | 1053 | ||
NM_001290655.1 | NP_001277584.1 | 9 | 1077 | |||
NM_172391.4 | NP_765979.3 | 9 | 1058 | |||
GenBank mRNA | AK033696.1 | - | 9 | 966 | ||
AK035941.1 | - | 9 | 831 | |||
AK093945.1 | - | 9 | 965 | |||
AK137116.1 | - | 9 | 969 | |||
AK154196.1 | - | 9 | 969 | |||
AK155265.1 | - | 9 | 969 | |||
BC038397.1 | - | 9 | 944 | 3676293 | ||
BC052829.1 | - | 9 | 995 | 30040034 | ||
BC055873.1 | - | 9 | 920 | |||
BC064012.1 | - | 9 | 878 | 30135289 |