Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3pxd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177364.3 | NP_796338.2 | 13 | 1459 | ||
GenBank mRNA | AB430861.1 | - | 13 | 1381 | ||
AK089330.1 | - | 13 | 1431 | |||
AK090119.1 | - | 13 | 1453 | |||
AK154898.1 | - | 13 | 1417 | |||
AK171384.1 | - | 13 | 1431 | |||
BC115711.1 | - | 13 | 1428 | |||
BC115764.1 | - | 13 | 1428 | |||
BC141306.1 | - | 13 | 1431 | 9055918 |