Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172945.2 | NP_766533.2 | 2 | 365 | ||
GenBank mRNA | BC151158.1 | - | 2 | 343 | 9088046 | |
BC172028.1 | - | 2 | 343 | |||
CA315262.1 | - | 1 | 51 | |||
CA329030.1 | - | 2 | 353 | |||
CJ131946.1 | - | 2 | 365 |