Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178743.3 | NP_848858.2 | 7 | 945 | ||
GenBank mRNA | AF345196.1 | - | 7 | 945 | ||
AK052202.1 | - | 2 | 1032 | |||
AK078545.1 | - | 6 | 2025 | |||
AK089182.1 | - | 7 | 925 | |||
BC038604.1 | - | 7 | 938 | |||
BC132493.1 | - | 2 | 813 | 40130770 | ||
BC137897.1 | - | 2 | 813 | 8860916 |