Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172951.3 | NP_766539.2 | 13 | 1187 | ||
GenBank mRNA | AF367760.1 | - | 13 | 1042 | ||
AK008673.1 | - | 13 | 1097 | |||
AK035310.1 | - | 13 | 1141 | |||
AK087560.1 | - | 13 | 1137 | |||
AK137345.1 | - | 13 | 1146 | |||
BC058204.1 | - | 12 | 949 | |||
BC138956.1 | - | 13 | 1101 | 8861978 | ||
BC145476.1 | - | 13 | 1096 |