Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002812.4 | NP_002803.2 | 3 | 570 | ||
GenBank mRNA | AK301771.1 | - | 3 | 581 | ||
AK308809.1 | - | 3 | 659 | |||
BC001164.1 | - | 3 | 321 | 3528096 | ||
BC065006.1 | - | 3 | 309 | 6055235 | ||
BI560993.1 | - | 3 | 554 | |||
BQ684248.1 | - | 3 | 333 | |||
BT019688.1 | - | 3 | 239 | |||
D38047.1 | - | 3 | 309 | |||
EU668354.1 | - | 3 | 570 |