Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gulo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178747.3 | NP_848862.1 | 4, 5 | 486 | ||
GenBank mRNA | AK077740.1 | - | 4, 5 | 487 | ||
AK167460.1 | - | 4, 5 | 475 | |||
AY453064.1 | - | 3, 4 | 323 | |||
BC019856.1 | - | 4, 5 | 433 | 5123684 | ||
BC028822.1 | - | 4, 5 | 442 | 5097681 | ||
BC028828.1 | - | 4, 5 | 445 | 5101228 | ||
BC034835.1 | - | 3, 4 | 242 |