Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081280.1 | NP_001074749.1 | 13 | 3105 | ||
NM_175547.3 | NP_780756.1 | 10 | 1028 | |||
GenBank mRNA | AK079766.1 | - | 10 | 952 | ||
AK084199.1 | - | 10 | 942 | |||
AK171971.1 | - | 12 | 4118 | |||
BC130223.1 | - | 10 | 2494 | |||
BC152543.1 | - | 10 | 2503 |