Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170978.1 | NP_001164449.1 | 12 | 1032 | ||
NM_172961.3 | NP_766549.2 | 12 | 1032 | |||
GenBank mRNA | AK036128.1 | - | 12 | 977 | ||
AK132282.1 | - | 12 | 981 | |||
AK165348.1 | - | 12 | 983 | |||
BC058079.1 | - | 12 | 972 | 6826741 | ||
BC058521.1 | - | 12 | 972 | 6825487 |