Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173440.2 | NP_775616.1 | 4 | 1043 | ||
GenBank mRNA | AF053062.1 | - | 2 | 691 | ||
AK036273.1 | - | 4 | 1044 | |||
AK156233.1 | - | 4 | 1022 | |||
AK157467.1 | - | 4 | 1022 | |||
BC060232.1 | - | 3 | 841 | 6852643 | ||
BC075638.1 | - | 4 | 920 |