Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caskin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027937.2 | NP_082213.2 | 17 | 1899 | ||
GenBank mRNA | AK129327.1 | - | 15 | 1462 | ||
BC059916.1 | - | 8 | 731 | |||
BC060720.1 | - | 17 | 1663 | |||
BC138443.1 | - | 15 | 1594 | 8861463 | ||
BC145281.1 | - | 15 | 1594 | 9053835 |